Order Entry
Sweden
Orders LinkContactUsLinkComponent
Pulvertratt

Pulvertratt

80 mm, 75 mm, 22 mm
Katalog # 221-0160
Leverantör:   VITLAB
Pulvertratt
Katalog # 221-0160
Leverantör:  VITLAB
Pulvertratt
Pulvertratt

80 mm, 75 mm, 22 mm

Price
75,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents

PP, translucent.

  • Very good chemical resistance
  • Very good temperature resistance
  • Autoclavable

Used for transferring powders. Also suitable for liquids.

Specifikationer
  • Tratt, y.d.:
    80 mm
  • Längd:
    75 mm
  • Rör, y.d.:
    22 mm