Gå till:
Product Details & Documents
Synonymer: 4,4'-Diaminodifenylsulfon
- CAS-nummer:80-08-0
- Formel:C₁₂H₁₂N₂O₂S
- MDL-nummer:MFCD00007887
- Molecular Weight:248.31 g/mol
- :M-Clarity™ quality level = MQ200
- EINECS:201-248-4
Specifikationer
Specifikationstestresultat
- Solubility:0.38 g/l (37 °C)
- Melting Point:172 - 175 °C
- Molar Mass:248.30 g/mol
- Bulk Density:250 - 350 kg/m3
- pH:5.5 - 7.5 (H2O, 20 °C) (saturated aqueous solution)
- Synonyms:DDS, 4,4'-Sulfonyldianiline, Bis(p-aminophenyl) sulfone
- Hill Formula:C12H12N2O2S
- Chemical Formula:[4-(NH2)C6H4]2SO2
Specifikationer
- Förpackningstyp:Glass bottle
- Förp.:5 g



