Inlet liners
Silanized Glass Wool
Katalog # PERC03300905
Leverantör: PerkinElmer
Gå till:
Product Details & Documents
Liners protect GC columns against column contamination. These glass split and splitless inlet liners promote full vaporisation of samples, minimise mass discrimination, and achieve accurate and reproducible results.
- Liners packed with wool help to trap non-volatile material
Specifikationer
- Beskrivning:Silanized Glass Wool
- Seq:09