Orders LinkContactUsLinkComponent
Dip slides
Dip slides
PCA / MacConkey agar No.3, 10
Katalog # OXOIDS0166A
Dip slides
Katalog # OXOIDS0166A
CAS-nummer:  
Dip slides
PCA / MacConkey agar No.3, 10
CAS-nummer:  
Price
637,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 10 Items
Product Details & Documents
Simple and safe for an unskilled operator to use, these dip slides are coated on both sides with a solid culture medium with the opportunity for different media to be carried on each side, making the process time- and cost-effective. This sterile dip slide can be inoculated in the ward or doctor’s surgery and then returned to its plastic container for easy transport back to the laboratory.
  • Make colony counting for the bacterial assessment simple
Documents
Specifikationer
Specifikationer
  • Förp.:
    10
  • Beskrivning:
    PCA / MacConkey agar No.3
Specifikationer