MicroWell™ MiniTrays, Nunc™
MiniTrays med 72 brunnar, Klar, 72, -
732-2750
PS, with lid, non sterile. The low profile design and 10 µl well volume make these plates ideal for serological applications.
- Limited adsorption of proteins
- Optical clarity allows use with inverted microscopes
- Lids have keyed tabs for proper orientation
- Large raised alphanumeric coordinates
- Unique surface treatment to maximise sample and reagent mixing
Colour: Clear.
Well dimensions (mm): Top 3,71, Bottom 1,27, Depth 1,91.
External dimensions (mm): 84×59 (with lid).
- Ant. brunnar:72
- Steril:-
- Färg:Klar
- Beskrivning:MiniTrays med 72 brunnar
- Cat. No.:732-2750
- Miljöcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact



