pH indicator strips, Whatman™
Universal indikatorpapper, remsor som inte blöder, 2,0 - 9,0
Leverantör: Whatman products (Cytiva)
Gå till:
Produktdetaljer och dokument
Universal indicator strips which have been impregnated with a mixture of several indicators. On contact with the sample solution they assume a particular colour. A check against the colour comparision table supplied allows the pH to be determined.
- Instant pH readings
- Accurate for a wide range of routine pH testing
- Convenient and portable for field use
Specifikationer
- pH-område:2,0 - 9,0
- Beskrivning:Universal indikatorpapper, remsor som inte blöder