Cryostorage boxes, Cryostore™
Katalog # 479-4012
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
[EN]VWR CRYOBOX 81PL 1-2ML GREEN 1 * 5 Items Katalog #479-2396
Product 479-2396 is similar to 479-4012 but manufacturer and specifications may vary.
Produktdetaljer och dokument
PC, coloured.
- Writable surfaces
- Available with four colours
- Printed with squared numbers
- Transparent cover allows to see the content of the box
These durable cryogenic storage boxes are designed to be used at temperatures between −196 °C and +121 °C. It can accommodate around 42×T310-10A Cryovials®.
Documents
SIMPT314-581B_TDS_EN
Datablad