Scoops
500 ml, 325 mm
Katalog # NICK9030
Leverantör: NICKEL ELECTRO
Gå till:
Produktdetaljer och dokument
Stainless steel scoops for collecting or measuring powders, pastes and granulates.
- Excellent thermal resistance, autoclavable
- Non magnetic, bright finish
- Highly polished surface makes cleaning easier
Specifikationer
- Kapacitet:500 ml
- Längd:325 mm