Dropping bottles, PP, Nalgene®
250 ml
Katalog # NALGDS2420-0250
Leverantör: Thermo Fisher Scientific
Dropping bottles, PP, Nalgene®
Katalog # NALGDS2420-0250
Leverantör: Thermo Fisher Scientific
Gå till:
Product Details & Documents
PPCO med droppfri stängning och lock av blått PP.
- Autoclavable even when full with tips uncapped
- Leakproof closure
- Integral non-drip closure with attached cap
Idealisk för sterila lösningar.
Specifikationer
- Kapacitet:250 ml
- Cat. No.:NALGDS2420-0250
- Miljöcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact