Dropping bottles, PP, Nalgene®
125 ml
Katalog # 216-4432
Leverantör: Thermo Fisher Scientific
Dropping bottles, PP, Nalgene®
Katalog # 216-4432
Leverantör: Thermo Fisher Scientific
Gå till:
Product Details & Documents
PPCO med droppfri stängning och lock av blått PP.
- Autoclavable even when full with tips uncapped
- Leakproof closure
- Integral non-drip closure with attached cap
Idealisk för sterila lösningar.
Specifikationer
- Kapacitet:125 ml
- Cat. No.:216-4432
- Miljöcertificering:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact