Absorptionsmedel
P200 minirulle, 3.75 l/minirulle
Katalog # 121-1005
Leverantör: 3M
Gå till:
Product Details & Documents
These absorption kits can be deployed wherever hazardous liquids are in use and can be used to contain and clean up a spill without delay.
- P110: för mindre spill, eller en sista upprensning av större spill, ark med 280×350 mm, kapacitet 0,25 l/ark
- P130: rulle för stora områden, kan kapas till rätt storlek, 330 mm×3 m, kapacitet 50 l/rulle
- P190: universalrulle, 330 mm × 3 m, kapacitet 72 l/rulle
- P200: minirulle, 75 mm×2 m, kapacitet 3,75 l/minirulle
- P300: kuddar 108×380 mm, kapacitet 2 l/kudde
- P-F2001: minirulle, kuddar, ark/rulle 12,5×1520 cm, absorption 119 l/pack
Documents
Specifikationer
- Beskrivning:P200 minirulle
- Seq:03
- Absorptionsförmåga:3.75 l/minirulle
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



