Spiralkylare
400 mm, 29/32 NS, 29/32 NS
Katalog # 201-3353
Leverantör: Lenz Laborglas
Gå till:
Product Details & Documents
DURAN®, borosilikatglas.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
For cooling the condensate. Water supply via glass tubing connectors with 11 mm external Ø.
Documents
BSE TSE Glass Columns
BSE/TSE-certifikat
Specifikationer
- Kärna:29/32 NS
- Mantellängd:400 mm
- NS-hylsa:29/32 NS
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



