Orders LinkContactUsLinkComponent
Multiple adapters
Multiple adapters
14/23 NS, 14/23 NS
Katalog # 271-2221
Leverantör:   Lenz Laborglas
Multiple adapters
Katalog # 271-2221
Leverantör:  Lenz Laborglas
Multiple adapters
Multiple adapters
14/23 NS, 14/23 NS
Price
835,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Produktdetaljer och dokument
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With one standard ground NS14/23 or NS 29/32 socket and two standard ground NS 14/23 or NS 29/32 cons. Side neck descending angle of 75°.
Dokument
Specifikationer
  • Kärna:
    14/23 NS
  • NS-hylsa:
    14/23 NS