Dropptratt
1000 ml, 29/32 NS, 29/32 NS, 20 ml
Katalog # 201-1129
Leverantör: Lenz Laborglas
Gå till:
Product Details & Documents
DURAN®, borosilicate glass, clear, with PTFE key.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Cylindrical shape. With standard ground joint cone and socket. Clearly visible white divisions for approximate volume reading.
DIN ISO 4800
: DIN ISO 4800
Specifikationer
- Kärna:29/32 NS
- NS-hylsa:29/32 NS
- Kapacitet:1000 ml
- Indelning:20 ml



