Dropptratt
500 ml, 29/32 NS, 29/32 NS, 10 ml
Katalog # 201-1128
Leverantör: Lenz Laborglas
Gå till:
Product Details & Documents
DURAN®, borosilicate glass, clear, with PTFE key.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Cylindrical shape. With standard ground joint cone and socket. Clearly visible white divisions for approximate volume reading.
DIN ISO 4800
Certifications: DIN ISO 4800
Documents
BSE TSE Glass Columns
BSE/TSE-certifikat
Specifikationer
- Kärna:29/32 NS
- NS-hylsa:29/32 NS
- Kapacitet:500 ml
- Indelning:10 ml



