Orders LinkContactUsLinkComponent
Kran
Kran
1,5 mm, 14,5 NS
Catalog # 201-1109
Supplier:   Lenz Laborglas
Kran
Catalog # 201-1109
Supplier:  Lenz Laborglas
Kran
Kran
1,5 mm, 14,5 NS
Price
975,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
DURAN®, borosilicate glass, with solid glass or PTFE key.
  • Utbytbar NS-krankik med T-rör
  • Gängad fastsättning
  • Täthet uppfyller DIN 12540-1
DIN 12554
Documents
Specifications
  • Hål Ø.:
    1,5 mm
  • Kran:
    14,5 NS