Orders LinkContactUsLinkComponent
Sprutflaskor, smal öppning
250 ml, 140 mm, 60 mm
  215-1365
 :   KARTELL
Sprutflaskor, smal öppning
  215-1365
 :  KARTELL
Sprutflaskor, smal öppning
Sprutflaskor, smal öppning
250 ml, 140 mm, 60 mm
81,00 kr
1 * 1 Items
LDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • Suitable for foodstuff
The stream of liquid is obtained by blowing through the straight tube. The direction of the stream of liquid can be controlled by hand.
  • Ø utv.:
    60 mm
  • Kapacitet:
    250 ml
  • Höjd:
    140 mm