Sprutflaskor, smal öppning
250 ml, 140 mm, 60 mm
215-1365
: KARTELL
LDPE, translucent.
- Good chemical resistance
- Good temperature resistance
- Suitable for foodstuff
The stream of liquid is obtained by blowing through the straight tube. The direction of the stream of liquid can be controlled by hand.
- Ø utv.:60 mm
- Kapacitet:250 ml
- Höjd:140 mm