Vägflaska, PP
360 ml, 70×118 mm
Katalog # 611-0105
Leverantör: KARTELL
Gå till:
Produktdetaljer och dokument
PP with lid.
- Very good chemical resistance
- Very good temperature resistance
Cylindrical shape, with knobbed lid.
Dokument
Specifikationer
- Diam×H:70×118 mm
- Kapacitet:360 ml