Vägflaska, PP
30 ml, 49×29 mm
Katalog # 611-9403
Leverantör: KARTELL
Gå till:
Product Details & Documents
PP with lid.
- Very good chemical resistance
- Very good temperature resistance
Cylindrical shape, with knobbed lid.
Documents
Specifikationer
- Diam×H:49×29 mm
- Kapacitet:30 ml