Drogtest kit
LSD reagens
Gå till:
Discontinued
Product Details & Documents
This kit provides a convenient means of screening suspect material for restricted drugs. The scheme and reagents were developed at the laboratory of the government chemist. Each kit provides sufficient reagents for at least 200 tests.
- Ett kit består av:
- 20 (4 lådor om 5 st, ampuller 0,2 ml) ampuller med markis reagens
- 1×10 ml kokain reagens
- 1×10 ml barbiturat reagens
- 1×10 ml LSD reagens
- 1×20 ml cannabis reagens 1
- 1×10 ml cannabis reagens 2
- 1×5 g cannabis special fast reagens
- 1x20 g neutralisatorer
- 4 lådor med Whatman filterpapper
- 1 microspatel· 20 ampull brytare· 25 engångsspatlar· Pincett· 4 urglas· Bruksanvisning
Documents
Certificates
Sök efter LOT-nummerSDS_SV_FI_VWRC32161 SDS_SV_FI_VWRC32161.pdf SDS_SV_FI_VWRC32162.pdf SDS_SV_FI_VWRC32163 SDS_SV_FI_VWRC32163.pdf
SDS
More
spec_VWRC321602G.pdf
Produktspecifikation
Specifikationer
- Beskrivning:LSD reagens
- Seq:06