Orders LinkContactUsLinkComponent
Drogtest kit
Drogtest kit
LSD reagens
Katalog # 321632M
Drogtest kit
Katalog # 321632M
Leverantör:  
Drogtest kit
Drogtest kit
LSD reagens
Discontinued
Product Details & Documents
This kit provides a convenient means of screening suspect material for restricted drugs. The scheme and reagents were developed at the laboratory of the government chemist. Each kit provides sufficient reagents for at least 200 tests.
  • Ett kit består av:
  • 20 (4 lådor om 5 st, ampuller 0,2 ml) ampuller med markis reagens
  • 1×10 ml kokain reagens
  • 1×10 ml barbiturat reagens
  • 1×10 ml LSD reagens
  • 1×20 ml cannabis reagens 1
  • 1×10 ml cannabis reagens 2
  • 1×5 g cannabis special fast reagens
  • 1x20 g neutralisatorer
  • 4 lådor med Whatman filterpapper
  • 1 microspatel· 20 ampull brytare· 25 engångsspatlar· Pincett· 4 urglas· Bruksanvisning
Specifikationer
  • Beskrivning:
    LSD reagens
  • Seq:
    06