Orders LinkContactUsLinkComponent
Drogtest kit
Drogtest kit
BDH drogtest, kit (kokain, barbiturater, LSD, cannabis)
Katalog # 321481P
Leverantör:   VWR Chemicals
Drogtest kit
Katalog # 321481P
Leverantör:  VWR Chemicals
Drogtest kit
Drogtest kit
BDH drogtest, kit (kokain, barbiturater, LSD, cannabis)
Price
12 875,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
This kit provides a convenient means of screening suspect material for restricted drugs. The scheme and reagents were developed at the laboratory of the government chemist. Each kit provides sufficient reagents for at least 200 tests.
  • Ett kit består av:
  • 20 (4 lådor om 5 st, ampuller 0,2 ml) ampuller med markis reagens
  • 1×10 ml kokain reagens
  • 1×10 ml barbiturat reagens
  • 1×10 ml LSD reagens
  • 1×20 ml cannabis reagens 1
  • 1×10 ml cannabis reagens 2
  • 1×5 g cannabis special fast reagens
  • 1x20 g neutralisatorer
  • 4 lådor med Whatman filterpapper
  • 1 microspatel· 20 ampull brytare· 25 engångsspatlar· Pincett· 4 urglas· Bruksanvisning
Specifikationer
  • Beskrivning:
    BDH drogtest, kit (kokain, barbiturater, LSD, cannabis)
  • Seq:
    01