Stopcocks, straight through, with tip
PP
Katalog # 661-1183
Leverantör: COWIE
Gå till:
Produktdetaljer och dokument
Made of PTFE.
- Available with PP or glass tip
- Very good chemical resistance
- Very good temperature resistance
Specifikationer
- Beskrivning:PP