Pipette tips, microvolume, Corning®
Rund spets, 0,1 - 10 µl, -, Transparent, Gilson, andra ultramikropipetter, Bulk; 10 000 spetsar/påse
Supplier: Corning
Jump to:
Product Details & Documents
These microvolume tips provide accurate, reliable performance and are designed for use with major brand ultra-micropipettors.
- All racked tips are certified RNase-/DNase-free and non pyrogenic
- Exceptional durability: all tip racks stand up to the demands of use with multi-channel pipettors and withstand repeated autoclaving cycles
Documents
Certificates
Search by Lot NumberSpecifications
- Förp.:Bulk; 10 000 spetsar/påse
- Steril:-
- Volym:0,1 - 10 µl
- Seq:02
- Tip style:Rund spets
- Spets färg:Transparent
- För användning med:Gilson, andra ultramikropipetter
- Cat. No.:613-4245
- Miljöcertificering:I-RECs, ISO 14001 - Reynosa
- Environmentally Preferable:Low Manufacturing Impact