Fluorbensen 99%
Glass bottle for solids, 500 g
: Phenylfluoride
- :462-06-6
- :C₆H₅F
- :MFCD00000280
- :96.1 g/mol
- :13,04197
- :85 °C (760 mmHg)
- :–42 °C
- :1,02 g/cm³
- :–15 °C
- :Omgivande
- :207-321-7
- :2387
- :3,II
Documents
ACRO11934
SDS
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4643 to 1.4663 (20°C, 589 nm)
- Water:≤0.1 % (K.F.)
- GC:≥98.5 %
- Förpackningstyp:Glass bottle for solids
- Förp.:500 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



