Basic Violet 3 90+% ACS
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 90+% CAS No.: 548-62-9 Molecular Formula: C25H30ClN3 Formula Weight: 407.99 Physical Form: Powder Melting Point: ca 215° dec. Beilstein Reference: 4077708 Merck Reference: 14,4395 UN No.: 3077 MDL No.: MFCD00011750
Synonyms: 4-[4,4'-Bis(dimetylamino) benzhydryliden]cyklohexa-2,5-dien-1-ylidendimetylammoniumklorid, CI 42555
- CAS Number:548-62-9
- Formula:C₂₅H₃₀ClN₃
- MDL Number:MFCD00011750
- Molecular Weight:407.99 g/mol
- Merck Index:14,4395
- Melting Pt:∼ 215 °C (dec.)
- Density:1,19 g/cm³ (20 °C)
- Storage Temperature:Omgivande
- EINECS:208-953-6
- UN:3077
- ADR:9,III
Documents
Specifications
Specifications
- Förpackningstyp:Glass bottle for solids
- Förp.:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



