Order Entry
Sweden
Orders LinkContactUsLinkComponent
HEPES BUFFER 1M IN NORMAL SALINE 1 * 100 ml

HEPES BUFFER 1M IN NORMAL SALINE 1 * 100 ml

Katalog # LONZ17-737E
HEPES BUFFER 1M IN NORMAL SALINE 1 * 100 ml
Katalog # LONZ17-737E
Leverantör:  
HEPES BUFFER 1M IN NORMAL SALINE 1 * 100 ml
Discontinued
Produkten har utgått. Men vi hittade liknande alternativa produkter.
Alternativ
HEPES BUFFER 1 M 1 * 100 mL Katalog #L0180-100

Produkten L0180-100 liknar LONZ17-737E men kan ha en annan tillverkare och/eller förpackning.

HEPES BUFFER 1 M 1 * 100 mL Katalog #L0180-100

Produkten L0180-100 liknar LONZ17-737E men kan ha en annan tillverkare och/eller förpackning.

HEPES, Free Acid, ULTROL Grade, 1M Solut 1 * 100 mL Katalog #375368-100

Produkten 375368-100 liknar LONZ17-737E men kan ha en annan tillverkare och/eller förpackning.

Product Details & Documents

Documents

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.