Order Entry
Sweden
Orders LinkContactUsLinkComponent
Trypsin

Trypsin

500 mL
Catalog # LONZ17-161F
Supplier:   LONZA
Trypsin
Catalog # LONZ17-161F
Supplier:  LONZA
CAS Number:  
Trypsin

500 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 500 mL
Product Details & Documents

Trypsin is a clear, colourless-yellow, highly active porcine enzyme preparation. It is generally used at a working concentration of 0,25%, prepared in a balanced salt solution at a pH of 7,6 to 7,8 without calcium or magnesium. It is used to remove cells from the culture vessel surface into suspension.

Trypsin-Versene® or Trypsin-EDTA are combined reagents used to remove cells from the culture vessel surface into suspension. The EDTA, a chelator of divalent cations, is added to trypsin solutions to enhance activity and to remove the calcium and magnesium from the cell surface.

Versene® (EDTA) is a chelating agent used to lift some cell types from the culture vessel surface into suspension. It is used as a wash prior to the addition of a trypsin product.

Versene® (EDTA) is a chelating agent used to lift some cell types from the culture vessel surface into suspension. It is used as a wash prior to the addition of a trypsin product.

Specifications

Specifications

  • Förp.:
    500 mL
  • Enzymnamn:
    Trypsin
  • Cat. No.:
    LONZ17-161F
  • Koncentration:
    1X

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.