Order Entry
Sweden
Orders LinkContactUsLinkComponent
Trypsin

Trypsin

500 ml
Katalog # LONZ17-160F
Trypsin
Katalog # LONZ17-160F
Leverantör:  
CAS-nummer:  
Trypsin

500 ml

CAS-nummer:  
Discontinued
Produkten har utgått. Men vi hittade liknande alternativa produkter.
Alternativ
DPBS WITH CA-MG W/O PHENOL RED 1 * 500 mL Katalog #HYCLSH30264.01

Produkten HYCLSH30264.01 liknar LONZ17-160F men tillverkare och specifikationer kan variera.

Product Details & Documents

Trypsin is a clear, colourless-yellow, highly active porcine enzyme preparation. It is generally used at a working concentration of 0,25%, prepared in a balanced salt solution at a pH of 7,6 to 7,8 without calcium or magnesium. It is used to remove cells from the culture vessel surface into suspension.

Trypsin-Versene® or Trypsin-EDTA are combined reagents used to remove cells from the culture vessel surface into suspension. The EDTA, a chelator of divalent cations, is added to trypsin solutions to enhance activity and to remove the calcium and magnesium from the cell surface.

Versene® (EDTA) is a chelating agent used to lift some cell types from the culture vessel surface into suspension. It is used as a wash prior to the addition of a trypsin product.

Versene® (EDTA) is a chelating agent used to lift some cell types from the culture vessel surface into suspension. It is used as a wash prior to the addition of a trypsin product.

Specifikationer

Specifikationer

  • Förp.:
    500 ml
  • Enzymnamn:
    Trypsin
  • Cat. No.:
    LONZ17-160F
  • Koncentration:
    1:250 (10X)

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.