Kran, sidoställd
Spare PTFE key for 25 ml and 50 ml burette capacity, Automatic burette, Pellet pattern
Katalog # BRND81167
Leverantör: Brand
Gå till:
Produktdetaljer och dokument
Borosilicate glass 5,4, amber.
- Interchangeable key
- With glass or PTFE key
- Key with retention device
Stopcock housing with burette tip and key.
Specifikationer
- För:Automatic burette, Pellet pattern
- Beskrivning:Spare PTFE key for 25 ml and 50 ml burette capacity
- Seq:06