Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

Byrett, sidoböjd kran, Schellbach, BLAUBRAND®

25 ml, 0,05 ml, 0,03 ± ml
  612-9942
 :   Brand
Byrett, sidoböjd kran, Schellbach, BLAUBRAND®
  612-9942
 :  Brand
Byrett, sidoböjd kran, Schellbach, BLAUBRAND®
Byrett, sidoböjd kran, Schellbach, BLAUBRAND®

25 ml, 0,05 ml, 0,03 ± ml

 
1 575,00 kr
   
1 * 1 Items
 

Borosilicate glass 3.3, class AS, DE-M marking.

  • Calibrated to deliver 'TD, Ex'
  • Schellbach stripe, ring marks at major graduations
  • Marks and inscriptions in high contrast blue enamel represents the optimum combination of resistance and readability

Length: 800 mm.

Längd: 800 mm

 : DIN EN ISO 385

 : Supplied with a batch certificate. Burettes are also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.

 
  • Kapacitet:
    25 ml
  • Indelning:
    0,05 ml
  • Tolerans:
    0,03 ± ml