Ammoniumformat 99%
Glass bottle for solids, 1 kg
Leverantör: Thermo Fisher Scientific
Gå till:
Product Details & Documents
Synonymer: Formic acid ammonium salt
- CAS-nummer:540-69-2
- Formel:HCO₂NH₄
- MDL-nummer:MFCD00013103
- Molecular Weight:63.06 g/mol
- Merck Index:13,00525
- Smält Pt:115…120 °C
- Densitet:1,28 g/cm³ (25 °C)
- Förvaringstemperatur:Omgivande
- EINECS:208-753-9
Documents
Specifikationer
Specifikationstestresultat
- Appearance:White Powder or crystals
- Infrared spectrum:Conforms
- Titration excess NaOH:≥98.5 %
- Chloride (Cl):≤0.005 %
- Sulfate (SO4):≤0.01 %
- Iron (Fe):≤0.002 %
- Heavy metals (as Pb):≤0.002 %
Specifikationer
- Förpackningstyp:Glass bottle for solids
- Förp.:1 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



