Instrumentskål med lock
300×150×70 mm
Katalog # 216-0019
Leverantör: BOCHEM
Gå till:
Product Details & Documents
Rostfritt stål, korrosionsbeständigt, 18/10.
- Special corner shape for fast, easy cleaning
- Polished electrolytically, anti-magnetic
- Good chemical resistance, very good thermal resistance
Med överfallande lock, stapelbar.
Specifikationer
- B×D×H:300×150×70 mm