Instrumentskål med lock
180×90×30 mm
Catalog # 216-0016
Supplier: BOCHEM
Jump to:
Product Details & Documents
Rostfritt stål, korrosionsbeständigt, 18/10.
- Special corner shape for fast, easy cleaning
- Polished electrolytically, anti-magnetic
- Good chemical resistance, very good thermal resistance
Med överfallande lock, stapelbar.
Specifications
- B×D×H:180×90×30 mm