Orders LinkContactUsLinkComponent
Instrumentskål med lock
180×90×30 mm
Catalog # 216-0016
Supplier:   BOCHEM
Instrumentskål med lock
Catalog # 216-0016
Supplier:  BOCHEM
Instrumentskål med lock
Instrumentskål med lock
180×90×30 mm
Price
960,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Rostfritt stål, korrosionsbeständigt, 18/10.
  • Special corner shape for fast, easy cleaning
  • Polished electrolytically, anti-magnetic
  • Good chemical resistance, very good thermal resistance
Med överfallande lock, stapelbar.
Specifications
  • B×D×H:
    180×90×30 mm