Stand bases
2,0 kg, 150×300×3 mm
Catalog # BOCH5002
Supplier: BOCHEM
Jump to:
Product Details & Documents
Stand bases made of stainless steel 18/10 with expanded, flat and heavy feet for stability.
- With M 10 threaded hole (off-centre)
- One adjustable foot
- Anti-slip rubber protection
Certifications: DIN 12892
Specifications
- B×D×H:150×300×3 mm
- Vikt:2,0 kg