Wire baskets, autoclavable, Poxygrid®
510×280×90 mm
Katalog # BELAH167670000
Leverantör: Bel-Art Products, a Part of SP
Wire baskets, autoclavable, Poxygrid®
Katalog # BELAH167670000
Leverantör: Bel-Art Products, a Part of SP
Gå till:
Produktdetaljer och dokument
These epoxy-coated, steel wire baskets are suitable for transporting and batch dipping glassware.
- Resistant to rust, corrosion, solvents, acids and alkalis
- Withstands a temperature range of −223 to +121 °C
Specifikationer
- B×D×H:510×280×90 mm