Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TUBING, FEP, 1/8'' OD X 1.59 MM ID 1 * 3 m
JOURJR-T-6804-M3
:
VICI JOUR RESEARCH
TUBING, FEP, 1/8'' OD X 1.59 MM ID 1 * 3 m
JOURJR-T-6804-M3
:
VICI JOUR RESEARCH