Orders LinkContactUsLinkComponent
Transfer capillaries
100 mm, 1,75 mm, 1,30
Catalog # HIRS9201710
Supplier:   HIRSCHMANN
Transfer capillaries
Catalog # HIRS9201710
Supplier:  HIRSCHMANN
Transfer capillaries
Transfer capillaries
100 mm, 1,75 mm, 1,30
Price
1 025,00 kr
Log in for availability
1 * 1.000 Items
Product Details & Documents
Klart glas, båda ändar öppna.
Specifications
  • Ø utv.:
    1,75 mm
  • Ø inr.:
    1,30
  • Längd:
    100 mm
  • Seq:
    25