Trattar, PP
110 mm, 10 mm, 75 mm
Katalog # HIRS7750105
Leverantör: HIRSCHMANN
Gå till:
Product Details & Documents
Funnels, made of PP, translucent. With smooth interior surfaces.
- Excellent chemical resistance
- Autoclavable
- Suitable for use with standard filter paper
Specifikationer
- Tratt, y.d.:75 mm
- Rör, y.d.:10 mm
- Rörlängd:110 mm