Trattar, PP
47 mm, 5 mm, 30 mm
Katalog # HIRS7750102
Leverantör: HIRSCHMANN
Gå till:
Product Details & Documents
Funnels, made of PP, translucent. With smooth interior surfaces.
- Excellent chemical resistance
- Autoclavable
- Suitable for use with standard filter paper
Specifikationer
- Tratt, y.d.:30 mm
- Rör, y.d.:5 mm
- Rörlängd:47 mm