Orders LinkContactUsLinkComponent
Trattar, PP
47 mm, 2 mm, 30 mm
Katalog # HIRS7750101
Leverantör:   HIRSCHMANN
Trattar, PP
Katalog # HIRS7750101
Leverantör:  HIRSCHMANN
Trattar, PP
Trattar, PP
47 mm, 2 mm, 30 mm
Price
445,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 24 Items
Product Details & Documents
Funnels, made of PP, translucent. With smooth interior surfaces.
  • Excellent chemical resistance
  • Autoclavable
  • Suitable for use with standard filter paper
Specifikationer
  • Tratt, y.d.:
    30 mm
  • Rör, y.d.:
    2 mm
  • Rörlängd:
    47 mm