Dissolved oxygen bottles
Flaskor för löst syre enligt Winkler, 200-300 ml, 19/26
Katalog # 215-1245
Leverantör: HIRSCHMANN
Gå till:
Product Details & Documents
Oxygen bottles according to Winkler, with white inscription label.
- The adjusted capacity is individually marked on the bottle
- A unique serial number is engraved on the bottle and stopper
Specifikationer
- Kapacitet:200-300 ml
- Beskrivning:Flaskor för löst syre enligt Winkler
- NS-propp:19/26