Micro pipettes, disposable, with ring mark
1/2/3/4/5 µl, Vit, Fem markeringar
Catalog # HECH40555003
Supplier: GLASWARENFABRIK KARL HECHT
Micro pipettes, disposable, with ring mark
Catalog # HECH40555003
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Disposable micro pipettes with ring mark.
- Calibrated to contain 'In'
- Colour-coded
Certifications: DIN ISO 7550
Specifications
- Färgkod:Vit
- Typ:Fem markeringar
- Kapacitet:1/2/3/4/5 µl
- Seq:01