Micro pipettes, disposable, with ring mark
25 µl, 2 × Vit, En markering
Katalog # HECH40555015
Leverantör: GLASWARENFABRIK KARL HECHT
Micro pipettes, disposable, with ring mark
Katalog # HECH40555015
Leverantör: GLASWARENFABRIK KARL HECHT
Gå till:
Product Details & Documents
Disposable micro pipettes with ring mark.
- Calibrated to contain 'In'
- Colour-coded
Certifications: DIN ISO 7550
Specifikationer
- Färgkod:2 × Vit
- Typ:En markering
- Kapacitet:25 µl
- Seq:02