Micro pipettes, disposable, with ring mark
100 µl, Blå, En markering
Catalog # HECH40555027
Supplier: GLASWARENFABRIK KARL HECHT
Micro pipettes, disposable, with ring mark
Catalog # HECH40555027
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Disposable micro pipettes with ring mark.
- Calibrated to contain 'In'
- Colour-coded
Certifications: DIN ISO 7550
Specifications
- Färgkod:Blå
- Typ:En markering
- Kapacitet:100 µl
- Seq:05