Buffert, bruksfärdig
Technical buffer TPL-7, 7,00, Plastic dosing bottle, 250 mL
Leverantör: WTW
Gå till:
Product Details & Documents
Every bottle has its own lot reference with the expiration date.
- Shelf life up to 3 years (unopened)
- Reference temperature at 25 °C
- Economic consumption
- Easiest handling by use of the dosing vessel
- One litre container with a separate compartment for the sample
Accuracy is ±0,03 pH
Certifications: Traceable to NIST.
Specifikationer
Specifikationer
- Förpackningstyp:Plastic dosing bottle
- Förp.:250 mL
- Beskrivning:Technical buffer TPL-7
- pH-värde:7,00



