Intelsius® Pathopouch
95 kPa pouch for pathology, A4, 308×230 mm
Catalog # DGINBP003
New Product
Supplier: DGP INTELSIUS
Jump to:
Product Details & Documents
The Intelsius® Pathopouch range are 95 kPa flexible pressure vessels designed to meet with transport regulations.
95 kPa plastic secondary packaging
- Outer pouch for paperwork
- Leak-proof
They are leak-proof and suitable to hold a wide range of primary containers, such as blood vials, specimen containers, swabs, blood collection bags and medical devices.
Certifications: ISTA Certified Class 6.2 Category A & B
Specifications
- Beskrivning:95 kPa pouch for pathology
- L×B:308×230 mm
- Seq:1
- Storlek:A4