[EN]SOCKET PANEL 1 * 1 Items
Katalog # WTWAREP6-00260014
New Product
Leverantör: WTW
Gå till:
Produktdetaljer och dokument
Specifikationer
- Western blot:Yes
- Clonality:Monoclonal
- Flow cytometry:Yes
- Antibody type:Primary
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Storage temperature:Cool
- Host:Mouse
- Shipping temperature:Ice
- Antigen name:Retinal Pigment Epithelium-specific 65 kDa protein
- Cat. No.:NOVUNB100-355SS
- Reactivity:Human
- Gene ID:6121
- Antigen synonyms:sRPE65|all-trans-retinyl-palmitate hydrolase|retinol isomerase|retinoid isomerohydrolase|retinitis pigmentosa 20 (autosomal recessive)|retinal pigment epithelium-specific 65 kDa protein|RBP-binding membrane protein|p63|LCA2|RP20|rd12|mRPE65
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:RPE65
- Clone:401.8B11.3D9