Jars, without Cap
PP, 250 ml, —, —, 83,4 mm
Katalog # RIXI4-1101-0250-65
New Product
Leverantör: BERLIN PACKAGING RIXIUS GMBH
Jars, without Cap
Katalog # RIXI4-1101-0250-65
Leverantör: BERLIN PACKAGING RIXIUS GMBH
New Product
Gå till:
Product Details & Documents
These slip lid jars are made of white PP.
- Very good chemical resistance
- Very good temperature resistance
Ordering information: Caps need to be ordered seperately.
Caution: *Produkten/produkterna på denna sida ligger utanför tillämpningsområdet för EU -direktivet 2019/904
Specifikationer
- Ø utv.:—
- Typ:PP
- Kapacitet:250 ml
- Höjd:83,4 mm
- Seq:4
- Hals, innerdiam.:—