Digital Microscopes, Edge
Product Details & Documents
The Dino-Lite Edge models provide superior image quality and a high level of flexibility due to the adaptable cap design. The built-in adjustable polarizer reduces glare and reflection on shiny objects.
This model can work with two magnification rates while maintaining the same working distance, making this the ideal model for macro view applications with a larger field of view and a zoom function to reveal smaller details. Different caps are supplied with the microscope making the Dino-Lite Edge the perfect choice for any professional application.
Specifikationer
- Förstoring:130 ~ 220×
- Modell:AM7515MZT2P
- Upplösning:2592×1944 pixels
- Beskrivning:Digital microscope, 5 MP with magnification lock
- Seq:6
- Belysning:8 white LEDs
- Arbetsavstånd:Standard
- Typ:Handheld digital microscope
- Cat. No.:IDCPAM7515MZT2P
- Datorkrav:Windows 7, 8, 10, 11, MacOS 10.14 and up
- Vikt:140 g
- Bildhastighet:Up to 30 fps
- Givare:CMOS
- Kamera:Digital camera, 5 MP
- Programvara:DinoCapture 2,0 (Windows), DinoXcope (Mac OS)