Bromophenol Red Indicator biological stain
Bottle
Katalog # APOSOR1012563-5G
New Product
Leverantör: Apollo Scientific
Bromophenol Red Indicator biological stain
Katalog # APOSOR1012563-5G
Leverantör: Apollo Scientific
New Product
CAS-nummer: 2800-80-8
Gå till:
Product Details & Documents
Usage: Organic building block/ reagent.
Synonymer: 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-bromophenol] S,S-dioxide
- CAS-nummer:2800-80-8
- Formel:C₁₉H₁₂Br₂O₅S
- MDL-nummer:MFCD00023014
- Molecular Weight:512,17 g/mol
Specifikationer
Specifikationer
- Förpackningstyp:Bottle